missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ accumet™ Waterproof AP72 pH/mV/Temperature Meter Kit
Shop All Fisherbrand Products
Click to view available options
Range (pH):
-2 to 16
Unit Size:
Each
Description
Includes
Meter Kit package includes meter, pH/ATC electrode, pH calibration buffers, sample bottle, and hard plastic carrying case.
- Ergonomic design fits comfortably in the hand
- Electrode bracket positions electrode for one-handed operation
- Advanced set-up mode lets user customize meter parameters and check electrode slope and offset to assure electrode accuracy
- Read pH, absolute mV or relative mV and temperature
- Accuracy of ±0.01pH and ±0.02mV
- Manual or automatic temperature control
- Five-point push-button pH calibration and automatic buffer recognition (pH 1.68, 4.01, 7.01, 10.01, 12.45)
- HOLD function, Ready indicator and automatic shut-off function
- Read pH, mV or relative mV and temperature in °C or °F
- Memory allows storage and recall of up to 50 readings with corresponding temperatures
Real-time clock that stamps stored data and calibration data with date and time
Six-point push-button pH calibration and automatic buffer recognition
Selectable buffer sets (NIST™, DIN and Standard US)
Certifications
ISO 9001:2000, CE
Warranty
Warranted to the end user to be free from defects in materials and workmanship under normal use for a period of three years.
Specifications
Specifications
| Includes | Meter, pH/ATC electrode, pH calibration buffers, sample bottle, hard Plastic carrying case |
| Accuracy (Temperature) | ±0.5°C |
| Accuracy (mV) | ±0.2 from 0 to ±400, ±2 beyond 200 |
| Accuracy (pH) | ±0.01 |
| For Use With (Application) | pH, mV, Temperature |
| Range (pH) | -2 to 16 |
| Resolution (pH) | 0.001 |
| Range (mV) | 0 to ±2000 |
| Resolution (mV) | 0.1 from 0 to ±400, 1 beyondmV |
| Calibration Points | 5 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction