missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Includes
Includes meter, batteries and instructions.
- Ergonomic design fits comfortably in the hand
- Electrode bracket positions electrode for one-handed operation
- Advanced set-up mode lets user customize meter parameters and check electrode slope and offset to assure electrode accuracy
- Read pH, absolute mV or relative mV and temperature
- Accuracy of ±0.01pH and ±0.02mV
- Manual or automatic temperature control
- Five-point push-button pH calibration and automatic buffer recognition (pH 1.68, 4.01, 7.01, 10.01, 12.45)
- HOLD function, Ready indicator and automatic shut-off function
- Read pH, mV or relative mV and temperature in °C
- Memory allows storage and recall of up to 16 readings with corresponding temperatures
Certifications
ISO 9001:2000, CE
Warranty
Warranted to the end user to be free from defects in materials and workmanship under normal use for a period of three years.
Specifications
Specifications
| Includes | Meter, batteries and instructions |
| Accuracy (Temperature) | ±0.5°C |
| Accuracy (mV) | ±0.2 from 0 to ±400, ±2 beyond 200 |
| Accuracy (pH) | ±0.01 |
| For Use With (Application) | pH, mV, Temperature |
| Range (pH) | -2 to 16 |
| Resolution (pH) | 0.001 |
| Range (mV) | 0 to ±2000 |
| Resolution (mV) | 0.1 from 0 to ±400, 1 beyondmV |
| Calibration Points | 5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?