missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ Temperature-Compensated Two-Cell Conductivity Probes, Mercury-Free

Product Code. 11550164 Shop All Fisherbrand Products
Change view
Click to view available options
Cell Constant:
1 cm-1
10 cm-1
Measurement Ranges:
1 to 200 mS/cm
10.0 to 2000 μS/cm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cell Constant Measurement Ranges unitSize
11550164 1 cm-1 10.0 to 2000 μS/cm Each
11530164 10 cm-1 1 to 200 mS/cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11550164

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

8-pin connectors

For use on accumet™ Excel (XL) series benchtop meters.

For use on accumet™ Excel (XL) series benchtop meters

  • Immersion-type cell
  • Use in standard electrode holder: unique compact size is same as typical pH electrode
  • Built-in temperature sensors provide temperature-compensated readings
  • Epoxy body

TRUSTED_SUSTAINABILITY

Specifications

Cell Constant 1 cm-1
Connector Types 8-pin
For Use With (Equipment) AB200, XL20, XL200, XL30, XL50, XL500, XL60, XL600 Benchtop Meters
Type Conductivity Probe
Material Epoxy
Measurement Ranges 10.0 to 2000 μS/cm
Storage Method Immersed in deionized water
Probe 2 Cell Conductivity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.