missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ accumet™ Solid-State Half-Cell ISEs, Mercury-Free: Fluoride, US Standard Connector
Shop All Fisherbrand ProductsDescription
For fluoride determinations.
- pH range: 5-9
- Material: Epoxy
- Connector type: US Standard
Specifications
Specifications
| For Use With (Application) | Determination of Fluoride concentration in water/wastewater, stack gases, explosives, etching baths, bioresearch, beverages, detergents |
| Material | Epoxy |
| Connector Types | U.S. Standard |
| Type | Solid-State Half-Cell ISEs |
| pH Range | 5 to 9 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?