missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ Micro and Semimicro Size Glass Combination Electrodes - Mercury - Free

Product Code. 11550064 Shop All Fisherbrand Products
Change view
Click to view available options
Connector Types:
BNC
U.S. Standard
Length (Metric):
60 mm
100 mm
150 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Length (Metric) unitSize
11550064 U.S. Standard 150 mm Each
11590104 BNC 100 mm Each
11500114 BNC 150 mm Each
11540064 U.S. Standard 100 mm Each
11560064 U.S. Standard 60 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11550064
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Ideal for microwell plates, serum cups, cuvets, micro test tubes, NMR tubes and capillary tubes

Zero to 14pH range
TRUSTED_SUSTAINABILITY

Specifications

Connector Types U.S. Standard
Junction Single
Material Glass
pH Range 0 to 14
Temperature (Metric) Operating -5°C to +110°C
Type pH/ORP Combination Titration Electrode, Extra-Long Ag/AgCl
Filling Solution Type Saturated KCl with AgCl. KCl
For Use With (Application) Small samples
Internal Reference Ag/AgCl
Temperature (English) Operating 23°F to 230°F
Temperature (Metric) Range -5°C to +110°C
Tip Type Micro
Warranty 1 Year
Length (Metric) 150 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.