missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ accumet™ Liquid-Filled Polymer Body Combination Electrodes
Ideal for measurements in small vessels or with small samples
Brand: Fisherbrand™
Code : 43
Additional Details : Weight : 0.01000kg
Description
Standard pH Combination Electrodes
- Rugged polypropylene or epoxy body—ideal for handheld use with aqueous samples
- Endcap seal keeps membrane moist and ready
- 0 to 14pH range
- All have porous ceramic junction
- Polypropylene body models are available with single-junction calomel or Ag/AgCl reference
- Epoxy Body models are available with standard-size body and calomel reference or in flat-surface model with Ag/AgCl reference
Calomel model eliminates measurement problems and clogging in research with Tris buffers.
Specifications
BNC | |
Polypropylene | |
pH Combination Electrode | |
Ideal for handheld use with aqueous samples | |
-5°C to +80°C | |
106 mm | |
10 mm |
Single Ceramic | |
0 to 14 | |
SP138-500 | |
Calomel or Ag/AgCl | |
4 in. | |
0.4 in. |