missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ Liquid-Filled pH/ATC Epoxy Body Combination Electrodes - Mercury-Free

Product Code. 11356152 Shop All Fisherbrand Products
Click to view available options
Connector Types:
BNC/Mini-phone
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11356152

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Single-junction models with Ag/AgCl reference element

  • Rugged standard-size epoxy body
  • Single porous ceramic junction
  • Built-in ATC probe
  • BNC pH/ATC connectors

  • Rugged epoxy body for use in field or plant
  • Ideal for handheld use with aqueous samples

Order Info

  • For use with accumet Excel (XL) Series, accumet Research (AR) Series and accumet Basic Series (AB15 and AB15 Plus) benchtop meters
  • Endcap seal; keeps membrane moist and ready

TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Connector Types BNC/Mini-phone
Junction Single Ceramic
Material Epoxy
pH Range 0 to 14
Temperature (Metric) Operating -5°C to +80°C
Type Combination pH Electrode
Filling Solution Type SP135-500
For Use With (Application) Ideal for handheld use with aqueous samples
For Use With (Equipment) All Accumet XL, AR, and AB series benchtop meters
Internal Reference Ag/AgCl
Probe ATC
Temperature (English) Operating 23°F to 212°F
Temperature (Metric) Range -5°C to +80°C
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.