missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ Glass Body Calomel Reference Electrodes

Product Code. 11523975 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Application):
Biomedical, Tris
Junction:
Sleeve
Storage Requirements:
Packed dry (without electrolyte)
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Junction Storage Requirements unitSize
11523975 Biomedical, Tris Sleeve Packed dry (without electrolyte) Each
11533975 Biomedical, Tris - - Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Product Code. 11523975

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

  • Standard-size glass body
  • Contains mercury
  • -5° to +80°C
  • 0 to 14pH range

TRUSTED_SUSTAINABILITY

Specifications

Connector Types Pin
Junction Sleeve
Material Glass
pH Range 0 to 14
Temperature (Metric) Operating -5°C to +80°C
Filling Solution Type Mercury
For Use With (Application) Biomedical, Tris
Internal Reference Hg/Hg2Cl2 (Calomel)
Reference Electrolyte Calomel
Storage Requirements Packed dry (without electrolyte)
Temperature (English) Operating 23°F to 176°F
Temperature (Metric) Range 80°C Max.
Length (English) 5.75 in.
Length (English) Cable 30 in.
Length (Metric) 14.6 cm
Length (Metric) Cable 76.2 cm
Diameter (English) Body 0.37 in.
Diameter (Metric) Body 1 cm
Diameter (Metric) Cap 1.6 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.