missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ Epoxy Body pH/ATC Probe for Corning Meters - Mercury-Free

Product Code. 11500164 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Connector Types:
BNC
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Unit Size
11500164 BNC Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11500164
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Estimated Shipment: 15-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

With BNC connector and ATC connector for Corning Meters

  • Epoxy body for durability
  • pH combination electrode with integral ATC probe and temperature signal connector specifically designed for use with Corning meters
  • Liquid filled, single-junction Ag/AgCl reference
  • BNC pH connector
TRUSTED_SUSTAINABILITY

Specifications

Connector Types BNC
Junction Single
Material Epoxy
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 80°C
Type pH/ATC Combination Electrode
Filling Solution Type Liquid
For Use With (Application) ATC, pH
For Use With (Equipment) pH meter
Internal Reference Ag/AgCl
Probe ATC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.