Learn More
Fisher BioReagents™ OptiGrow™ Preweighed Media
Optimized for superior yield of bacterial culture
Brand: Fisher BioReagents™ BP1425-10P1
Code : 32
Additional Details : Weight : 0.40000kg
Description
Four classic media, in convenient, preweighed pouches.
- Preblended growth-promoting components save time and preparation costs
- Reduce formulation errors
- Each pouch contains enough material for 1 liter of liquid media
- No more weighing of dusty powders
- High quality—assured by stringent testing procedures
- LB is used for the culturing of E. coli in molecular biology procedures
- Terrific Broth is a buffered enriched medium used to obtain high yields of plasmid DNA from transformed E. coli
Order Info
Supplied as 10 pouches per box.
Specifications
Culturing of Escherichia coli | |
Pouch | |
10 Pouches |