Learn More
Fisher BioReagents™ OptiGrow™ Preweighed Media
Optimized for superior yield of bacterial culture
£66.69 - £87.24
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
11894674
|
Fisher BioReagents™
BP1427-10P1 |
LB Broth Lennox; 10 Pouches/Box |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11820715
|
Fisher BioReagents™
BP1426-10P1 |
LB Broth Miller; 10 Pouches/Box |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11835445
|
Fisher BioReagents™
BP1425-10P1 |
LB Agar Miller; 10 Pouches/Box |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11492964
|
Fisher BioReagents™
BP2468-10P1 |
Terrific Broth; 10 Pouches/Box |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Four classic media, in convenient, preweighed pouches.
- Preblended growth-promoting components save time and preparation costs
- Reduce formulation errors
- Each pouch contains enough material for 1 liter of liquid media
- No more weighing of dusty powders
- High quality—assured by stringent testing procedures
- LB is used for the culturing of E. coli in molecular biology procedures
- Terrific Broth is a buffered enriched medium used to obtain high yields of plasmid DNA from transformed E. coli
Order Info
Supplied as 10 pouches per box.
Specifications
Pouch | |
10 Pouches |