missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Fish (rainbow trout) S9 Fractions
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Liver S9 fractions prepared from a pool of rainbow trout (male or female, depending on lot).
Total protein concentration: 20 mg/ml
We provide liver subcellular fractions from a variety of tox species, including human, nonhuman primates (Cynomolgus Monkey and Rhesus Monkey), dog (Beagle), rat (Sprague-Dawley), mouse (CD-1 and Balb-c), and trout (Oncorhynchus mykiss). Custom species are available upon request.
For research use only. Not intended for human or animal therapeutic or diagnostic use.
Related Link
Learn more about our microsomes and subcellular fractions
Total protein concentration: 20 mg/ml
We provide liver subcellular fractions from a variety of tox species, including human, nonhuman primates (Cynomolgus Monkey and Rhesus Monkey), dog (Beagle), rat (Sprague-Dawley), mouse (CD-1 and Balb-c), and trout (Oncorhynchus mykiss). Custom species are available upon request.
For research use only. Not intended for human or animal therapeutic or diagnostic use.
Related Link
Learn more about our microsomes and subcellular fractions
Specifications
Specifications
| Concentration | 20 mg/mL |
| Description | Fish (Rainbow Trout) S9 Fractions, 1mL, Frozen, 20mg/mL Concentration, Male, Liver Tissue, Fish Species, S9 Fraction Sub-Cellular Localization, For Hepatocytes and Hepatic Models, Pharma and Biopharma, Target-Based ADME/Tox Assays |
| Product Type | Subcellular Fractions |
| Content And Storage | -60°C and -85°C |
| Donor Source | Pooled |
| Form | Frozen |
| For Use With (Application) | Hepatocytes and hepatic models, pharma and biopharma, target-based ADME/Tox assays |
| Quantity | 1 mL |
| Format | Tube |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction