missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FIP1/RCP Antibody (1A12B9D8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52546-0.025mg
This item is not returnable.
View return policy
Description
FIP1/RCP Monoclonal specifically detects FIP1/RCP in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications
| FIP1/RCP | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready | |
| DKFZp686E2214, EC 3.2.1.28, EC 6.1.1.7, FLJ22524, FLJ22622, MGC78448, NOEL1A, RAB11 coupling protein, RAB11 family interacting protein 1 (class I), rab11 family-interacting protein 1, Rab11-FIP1, Rab-coupling protein, Rab-interacting recycling protein, RCPRab effector protein | |
| Mouse | |
| 137.2 kDa | |
| 0.025 mg | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1A12B9D8 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| RAB11FIP1 | |
| Purified recombinant fragment of human RAB11FIP1 (AA: 130-271) expressed in E. Coli. | |
| Protein A or G purified | |
| RUO | |
| 80223 | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction