missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FIP1/RCP Antibody (1A12B9D8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£161.00 - £435.00
Specifications
| Antigen | FIP1/RCP |
|---|---|
| Clone | 1A12B9D8 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18207090
|
Novus Biologicals
NBP2-52546 |
0.1 mg |
£435.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18671388
|
Novus Biologicals
NBP2-52546-0.025mg |
0.025 mg |
£161.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FIP1/RCP Monoclonal specifically detects FIP1/RCP in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| FIP1/RCP | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| DKFZp686E2214, EC 3.2.1.28, EC 6.1.1.7, FLJ22524, FLJ22622, MGC78448, NOEL1A, RAB11 coupling protein, RAB11 family interacting protein 1 (class I), rab11 family-interacting protein 1, Rab11-FIP1, Rab-coupling protein, Rab-interacting recycling protein, RCPRab effector protein | |
| RAB11FIP1 | |
| IgG1 | |
| Protein A or G purified |
| 1A12B9D8 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| 80223 | |
| Purified recombinant fragment of human RAB11FIP1 (AA: 130-271) expressed in E. Coli. | |
| Primary | |
| 137.2 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title