Learn More
Thermo Scientific™ Finnpipette™ F1 Fixed Volume Single-Channel Pipettes
Protect samples from contamination with this manual pipette, featuring an adjustable finger rest and antimicrobial surface.
Brand: Thermo Scientific™ 4651060
Code : 04
Additional Details : Weight : 0.16000kg
Description
Protect samples from contamination with the Thermo Scientific™ Finnpipette™ F1 Fixed-Volume Pipette, featuring an adjustable finger rest, antimicrobial surface and super blow-out piston with volumes of 50μL and below. The Finnpipette F1 Fixed-Volume Pipette delivers light, smooth pipetting action with patented soft-touch tip ejection for maximum user comfort. The antimicrobial surface helps keep the pipette clean without the use of additional surface decontamination liquids. The entire pipette is resistant to UV light, reagents and moisture.
Features:- Antimicrobial surface: Silver (Ag) ions create a pipette surface that inhibits the growth of bacteria, microbes and fungus
- Adjustable finger rest: Finger rest can be adjusted 120° for optimal pipetting position
- Light, smooth pipetting action: Enables better results over longer pipetting periods
- Super blow-out piston with volumes of 50μL and below: Ensures delivery of micro-size drops
- Proprietary soft-touch tip ejection: Minimizes forces needed for tip ejection
Specifications
+/-0.4% | |
FinnPipette™ F1 | |
Fixed Volume Pipette | |
0.3% |
F1 Fixed Volume Single Channel | |
1 | |
250 μL |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.