missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Finnpipette™ F1 Fixed Volume Single-Channel Pipettes

Product Code. p-4530151
Change view
Click to view available options
Volume (Metric):
1 μL
10 μL
10,000 μL
100 μL
1000 μL
20 μL
200 μL
2000 μL
25 μL
250 μL
3000 μL
5 μL
50 μL
500 μL
5000 μL
Accuracy:
±0.40%
±0.70
+/-0.3%
+/-0.4%
+/-0.6%
+/-0.9%
+/-1.4%
+/-4%
Precision:
0.2%
0.3%
0.4%
0.5%
0.8%
1.4%
4%
Unit Size:
Each
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Accuracy Precision unitSize
11847401 1 μL +/-4% 4% Each
11857401 5 μL +/-1.4% 1.4% Each
11867401 10 μL +/-0.9% 0.8% Each
11877401 25 μL +/-0.6% 0.5% Each
11887401 50 μL +/-0.6% 0.4% Each
11897401 100 μL +/-0.4% 0.3% Each
11807411 250 μL +/-0.4% 0.3% Each
11817411 500 μL +/-0.3% 0.3% Each
11827411 1000 μL +/-0.3% 0.3% Each
11837411 2000 μL +/-0.3% 0.2% Each
11847411 3000 μL +/-0.3% 0.2% Each
10715515 5000 μL +/-0.3% 0.2% Each
10192644 10,000 μL +/-0.3% 0.2% Each
12336552 20 μL ±0.70 0.5% Each
12346552 200 μL ±0.40% 0.3% Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Product Code. 11847401 Supplier Thermo Scientific™ Supplier No. 4651000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Protect samples from contamination with this manual pipette, featuring an adjustable finger rest and antimicrobial surface.

Protect samples from contamination with the Thermo Scientific™ Finnpipette™ F1 Fixed-Volume Pipette, featuring an adjustable finger rest, antimicrobial surface and super blow-out piston with volumes of 50μL and below. The Finnpipette F1 Fixed-Volume Pipette delivers light, smooth pipetting action with patented soft-touch tip ejection for maximum user comfort. The antimicrobial surface helps keep the pipette clean without the use of additional surface decontamination liquids. The entire pipette is resistant to UV light, reagents and moisture.

Features:
  • Antimicrobial surface: Silver (Ag) ions create a pipette surface that inhibits the growth of bacteria, microbes and fungus
  • Adjustable finger rest: Finger rest can be adjusted 120° for optimal pipetting position
  • Light, smooth pipetting action: Enables better results over longer pipetting periods
  • Super blow-out piston with volumes of 50μL and below: Ensures delivery of micro-size drops
  • Proprietary soft-touch tip ejection: Minimizes forces needed for tip ejection
TRUSTED_SUSTAINABILITY

Specifications

Accuracy +/-4%
Description F1 Fixed Volume Single Channel
For Use With FinnTip™ Flex 10, 10, 20 Micro, 50 Micro
Model F1
Number of Channels 1
Type Fixed Volume Pipette
Volume (Metric) 1 μL
Precision 4%

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.