Learn More
Agilent BioTek Filters for ELx800 Universal Microplate Reader
Accessory for use with ELx800 Universal Microplate Readers.
Brand:  Agilent BioTek 7334562
Code : 04
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10257702
  • £433.00 / Each
Estimated Shipment: 29-04-2024
to see stock.
Description
Description
Accessory for use with ELx800 Universal Microplate Readers.
Specifications
Specifications
Filter
562 nm