missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Filter Funnel with Standard Joint

Filter Funnel with fused filter disc.

Brand:  ROBU VitraPOR™ 47012

Additional Details : Weight : 0.05600kg

 View more versions of this product

Product Code. 16863096

  • £74.10 / Pack of 2
Estimated Shipment: 16-05-2024
to see stock.
Add to basket

Description

Description

  • With fused filter disc
  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications

Specifications

5 mL
35 mm
2
Borosilicate Glass 3.3
NSK 10/19
2/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers