missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Filter Funnel

Filter funnel available in a buchner shape or a conical shape.

Brand:  ROBU VitraPOR™ 2125F

Additional Details : Weight : 0.27840kg

 View more versions of this product

Product Code. 16833036

  • £88.25 / Each
Estimated Shipment: 16-05-2024
to see stock.
Add to basket

Description

Description

  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications

Specifications

250 mL
Borosilicate Glass 3.3
Buchner Shape
Fine
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers