Learn More
Agilent BioTek Filter Cube, Customizable Filter Cube
For use with microplate readers and imagers
Brand:  Agilent BioTek 8040593
Additional Details : Weight : 0.50000kg
Product Code. 17190549
  • £1070.00 / Each
Estimated Shipment: 23-05-2024
to see stock.
Description
Description
  • Accessory for Agilent BioTek instruments range of microplate washers, readers and cell imagers
  • Use standard filters and dichroic mirrors to create custom cube
  • Single filter set and dichroic mirror easily configured using unique cube design
Specifications
Specifications
Microplate Readers and Imagers