Learn More
Agilent BioTek Filter Cube
For use with Synergy LX
Brand:  Agilent BioTek 1505006
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15998550
  • £1405.00 / 1 unit
Estimated Shipment: 22-05-2024
to see stock.
Specifications
Synergy LX
EX 360/40, EM 460/40, 400 nm mirror (Blue)