Learn More
Agilent BioTek Filter Cube
For use with Synergy LX
Brand:  Agilent BioTek 1505005
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15988550
  • £1405.00 / 1 unit
Estimated Shipment: 20-05-2024
to see stock.
Specifications
Synergy LX
EX 485/20, EM 528/20, 510 nm mirror (Green)