Learn More
Duran™ Filter Candle With Tube
The Filter Candle with Tube is suitable for the extraction of supernatant solutions and also for the separation of solids from a carrier gas. Available with porosities ranging from 1 to 4.
Brand:  Duran™ 258505209
Code : NEW
Additional Details : Weight : 0.09200kg
 View more versions of this product
Product Code. 10349990
  • £62.40 / Each
Estimated Shipment: 18-06-2024
to see stock.
Add to basket
Description
Description
  • Clear DURAN™ borosilicate glass 3.3
  • Conical filter
  • With tube
Specifications
Specifications
Borosilicate Glass (3.3)
2
24 x 65 mm (Filter)
20 mm (Tube)
40 to 100 μm
1 Ea.
200 mm (Tube)