Description
- Overcaps made from polyethylene
- Filter adapter caps have peel-away covers for inserting barbed disc filters for sterile venting with 0.2 μm membrane
- Sterlized by gamma irradiation
- Solid red overcap is used in closed systems or applications where venting is not necessary
Specifications
Specifications
| Material | Polyethylene |
| No. per Case | 100/Cs. |
| No. per Pack | 5/Pk. |
| Sterilization Method | Gamma Irradiation |
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.