missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Filamin A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£248.00 - £435.00
Specifications
| Antigen | Filamin A |
|---|---|
| Dilution | Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18471982
|
Novus Biologicals
NBP1-90284 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18487681
|
Novus Biologicals
NBP1-90284-25ul |
25 μL |
£248.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Filamin A Polyclonal antibody specifically detects Filamin A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Filamin A | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS (pH 7.2) and 40% Glycerol | |
| 2316 | |
| IgG | |
| Immunogen affinity purified |
| Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ABP-280DKFZp434P031, ABPX, actin binding protein 280, Actin-binding protein 280, Alpha-filamin, CVD1, EC 2.1.1.43, EC 6.3.4.4, Endothelial actin-binding protein, filamin A, alpha, filamin A, alpha (actin binding protein 280), Filamin-1, filamin-A, FLN, FLN1OPD, FLN-A, FMD, MNS, NHBP, Non-muscle filamin, OPD1, OPD2, XLVD, XMVD | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: IKFADQHVPGSPFSVKVTGEGRVKESITRRRRAPSVANVGSHCDLSLKIPEISIQDMTAQVTSPSGKTHEAEIVEGENHTYCIRFVPAEMGTHTVSVKYKGQHVPGSPFQFTV | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title