missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Filamin A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-90284
This item is not returnable.
View return policy
Description
Filamin A Polyclonal antibody specifically detects Filamin A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Filamin A | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| ABP-280DKFZp434P031, ABPX, actin binding protein 280, Actin-binding protein 280, Alpha-filamin, CVD1, EC 2.1.1.43, EC 6.3.4.4, Endothelial actin-binding protein, filamin A, alpha, filamin A, alpha (actin binding protein 280), Filamin-1, filamin-A, FLN, FLN1OPD, FLN-A, FMD, MNS, NHBP, Non-muscle filamin, OPD1, OPD2, XLVD, XMVD | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: IKFADQHVPGSPFSVKVTGEGRVKESITRRRRAPSVANVGSHCDLSLKIPEISIQDMTAQVTSPSGKTHEAEIVEGENHTYCIRFVPAEMGTHTVSVKYKGQHVPGSPFQFTV | |
| 0.1 mL | |
| Neuroscience | |
| 2316 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction