missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Filaggrin Antibody (FLG/1563), Novus Biologicals™
Mouse Monoclonal Antibody
£221.00 - £373.00
Specifications
| Antigen | Filaggrin |
|---|---|
| Clone | FLG/1563 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL, Protein Array |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18611039
|
Novus Biologicals
NBP2-75771-20ug |
20 μg |
£221.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18645747
|
Novus Biologicals
NBP2-75771-100ug |
100 μg |
£373.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Filaggrin Monoclonal antibody specifically detects Filaggrin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Protein ArraySpecifications
| Filaggrin | |
| 0.2 mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Cellular Markers | |
| 10 mM PBS with 0.05% BSA | |
| ATOD2, epidermal filaggrin, filaggrin, Profilaggrin | |
| Recombinant human Filaggrin protein fragment (around aa 998-1104) (exact sequence is proprietary) (Uniprot: P20930) | |
| Primary | |
| Store at 4°C. |
| FLG/1563 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P20930 | |
| 2312 | |
| IgG2b κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title