missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Filaggrin Antibody (FLG/1563), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-75771-100ug
This item is not returnable.
View return policy
Description
Filaggrin Monoclonal antibody specifically detects Filaggrin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
Specifications
| Filaggrin | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL, Protein Array | |
| P20930 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
| FLG/1563 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| ATOD2, epidermal filaggrin, filaggrin, Profilaggrin | |
| Recombinant human Filaggrin protein fragment (around aa 998-1104) (exact sequence is proprietary) (Uniprot: P20930) | |
| 100 μg | |
| Cell Biology, Cellular Markers | |
| 2312 | |
| Store at 4°C. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction