missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ficolin-2 Antibody (19) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-05042
This item is not returnable.
View return policy
Description
Ficolin-2 Monoclonal specifically detects Ficolin-2 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation.
Specifications
| Ficolin-2 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:4000 | |
| 37 kDa elastin-binding protein, Collagen/fibrinogen domain-containing protein 2, EBP-37ficolin (collagen/fibrinogen domain-containing lectin) 2 (hucolin), FCNLficolin B, ficolin (collagen/fibrinogen domain containing lectin) 2 (hucolin), ficolin-2, Ficolin-B, Ficolin-beta, Hucolin, L-ficolin, P35, Serum lectin p35 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 2220 | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 19 | |
| Unconjugated | |
| Q15485 | |
| FCN2 | |
| Purified procaryot expressed recombinant human L-Ficolin and full-length L-Ficolin (Ficolin-2) protein | |
| 0.1 mL | |
| Primary | |
| Reacts specifically with L-Ficolin. No cross reactivity is seen with M-Ficolin (Ficolin-1) and H-Ficolin (Ficolin-3). | |
| Store at 4C. Do not freeze. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction