missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ficolin-2 Antibody (19) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£156.00 - £367.00
Specifications
| Antigen | Ficolin-2 |
|---|---|
| Clone | 19 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:4000 |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18239842
|
Novus Biologicals
NBP1-05042-0.025ML |
0.025 mL |
£156.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18206300
|
Novus Biologicals
NBP1-05042 |
0.1 mL |
£367.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ficolin-2 Monoclonal specifically detects Ficolin-2 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
| Ficolin-2 | |
| 1.0 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| 37 kDa elastin-binding protein, Collagen/fibrinogen domain-containing protein 2, EBP-37ficolin (collagen/fibrinogen domain-containing lectin) 2 (hucolin), FCNLficolin B, ficolin (collagen/fibrinogen domain containing lectin) 2 (hucolin), ficolin-2, Ficolin-B, Ficolin-beta, Hucolin, L-ficolin, P35, Serum lectin p35 | |
| FCN2 | |
| IgG2a κ | |
| Protein A or G purified | |
| Reacts specifically with L-Ficolin. No cross reactivity is seen with M-Ficolin (Ficolin-1) and H-Ficolin (Ficolin-3). |
| 19 | |
| Western Blot 1:100-1:2000, ELISA 1:4000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q15485 | |
| 2220 | |
| Purified procaryot expressed recombinant human L-Ficolin and full-length L-Ficolin (Ficolin-2) protein | |
| Primary | |
| Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title