missing translation for 'onlineSavingsMsg'
Learn More

Fibulin 5 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2A5C8, Proteintech

Mouse Monoclonal Antibody

£135.00 - £349.00

Specifications

Antigen Fibulin 5
Clone 2A5C8
Concentration 1.2 mg/mL
Applications Western Blot, Immunofluorescence, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16817055
View Documents
Proteintech
60081-1-IG-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16807055
View Documents
Proteintech
60081-1-IG-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Essential for elastic fiber formation, is involved in the assembly of continuous elastin (ELN) polymer and promotes the interaction of microfibrils and ELN. Stabilizes and organizes elastic fibers in the skin, lung and vasculature. Promotes adhesion of endothelial cells through interaction of integrins and the RGD motif. Vascular ligand for integrin receptors which may play a role in vascular development and remodeling.
Specifications

Specifications

Fibulin 5
1.2 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
ARMD3, DANCE, EVEC, FBLN5, FIBL 5, fibulin 5, UP50, Urine p50 protein
Fbln5
IgG1
Caprylic Acid Precipitation
Antibody
2A5C8
Western Blot, Immunofluorescence, Immunocytochemistry
Unconjugated
Mouse
Human, Mouse, Pig, Rat
Q9UBX5, Q9WVH8, Q9WVH9
100155248, 10516, 23876, 29158
Fibulin 5 Fusion Protein Ag2859
Primary
-20°C
FBLN5
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.