missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibulin 5 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2A5C8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60081-1-IG-20UL
This item is not returnable.
View return policy
Description
Essential for elastic fiber formation, is involved in the assembly of continuous elastin (ELN) polymer and promotes the interaction of microfibrils and ELN. Stabilizes and organizes elastic fibers in the skin, lung and vasculature. Promotes adhesion of endothelial cells through interaction of integrins and the RGD motif. Vascular ligand for integrin receptors which may play a role in vascular development and remodeling.Specifications
| Fibulin 5 | |
| Monoclonal | |
| 1.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9UBX5, Q9WVH8, Q9WVH9 | |
| Fbln5 | |
| Fibulin 5 Fusion Protein Ag2859 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Pig, Rat | |
| Antibody | |
| IgG1 |
| Western Blot, Immunofluorescence, Immunocytochemistry | |
| 2A5C8 | |
| Unconjugated | |
| FBLN5 | |
| ARMD3, DANCE, EVEC, FBLN5, FIBL 5, fibulin 5, UP50, Urine p50 protein | |
| Mouse | |
| Caprylic Acid Precipitation | |
| RUO | |
| 100155248, 10516, 23876, 29158 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction