missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibrinopeptide B, Rabbit anti-Human, Clone: 800000, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81111-0.2MG
This item is not returnable.
View return policy
Description
Fibrin beta-chain Monoclonal specifically detects Fibrin beta-chain in Human samples. It is validated for Flow Cytometry, ELISA, In vivo assay.
Specifications
| Fibrinopeptide B | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA, In vivo assay | |
| fibrinogen beta chain, fibrinogen, B beta polypeptide, MGC104327, MGC120405 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| 8E5 binds to the amino-terminal region of the beta-chain of human fibrin at an epitope consiting of GHRPLDKC. Binding is possible in both the monomeric form and the polymeric form of the protein. This peptide sequence is found only in fibrin, and not in its precursor fibrinogen and so there is no cross-reactivity with fibrinogen. Fibrin is a non-globular protein which plays a role in the clotting of blood and circulates as the precursor fibrinogen. Polymerization occurs through the action of the protease thrombin, which results in the active fibrin. Fibrin polymerizes as an insoluble matrix and forms a clot along with platelets over a wound site. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG κ |
| ELISA | |
| 800000 | |
| Unconjugated | |
| PBS | |
| FGB | |
| 8E5 was prepared fusing Sp2/o myeloma cells with spleen cells derived from female BALB/c mice immunized with a synthetic beta-fibrin peptide seqeunce GHRPLDKC conjugated to MB-KLH (maleimidobenzoylated keyhole limpet hemocyanin). | |
| 0.2 mg | |
| Cell Cycle and Replication | |
| 2244 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction