missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FHOD3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£168.00
Specifications
| Antigen | FHOD3 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 15 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
Description
FHOD3 Polyclonal specifically detects FHOD3 in Human, Mouse, Bovine, Canine, Chicken, Equine, Opossum samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| FHOD3 | |
| Immunohistochemistry-Paraffin 15 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Bovine, Canine, Chicken, Equine, Opossum | |
| Q2V2M9 | |
| 80206 | |
| A portion of amino acids 1250-1300 of human FHOD3 (CRA-d) was used as the immunogen. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| 0.2 mL PBS and 0.05% BSA with 0.05% Sodium Azide | |
| FH1/FH2 domain-containing protein 3, FHOS2, formactin-2, Formactin2, formin homolog overexpressed in spleen 2, formin homology 2 domain containing 3, hFHOS2, KIAA1695 | |
| FHOD3 | |
| IgG | |
| Affinity Purified | |
| The amino acid sequence used as immunogen is 100% homologous in human (isoform 1 and isoform 2), rat, mouse, zebrafish, xenopus and bovine. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title