missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FHOD3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24544SS
This item is not returnable.
View return policy
Description
FHOD3 Polyclonal specifically detects FHOD3 in Human, Mouse, Bovine, Canine, Chicken, Equine, Opossum samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| FHOD3 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 mL PBS and 0.05% BSA with 0.05% Sodium Azide | |
| FH1/FH2 domain-containing protein 3, FHOS2, formactin-2, Formactin2, formin homolog overexpressed in spleen 2, formin homology 2 domain containing 3, hFHOS2, KIAA1695 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 80206 | |
| Human, Mouse, Bovine, Canine, Chicken, Equine, Opossum | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Immunohistochemistry-Paraffin 15 μg/mL | |
| Q2V2M9 | |
| FHOD3 | |
| A portion of amino acids 1250-1300 of human FHOD3 (CRA-d) was used as the immunogen. | |
| 0.025 mg | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human (isoform 1 and isoform 2), rat, mouse, zebrafish, xenopus and bovine. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction