missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFR3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£150.00
Specifications
| Antigen | FGFR3 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:250, Immunohistochemistry-Paraffin 1:250 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
FGFR3 Polyclonal specifically detects FGFR3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| FGFR3 | |
| Western Blot 1:1000, Immunohistochemistry 1:250, Immunohistochemistry-Paraffin 1:250 | |
| Polyclonal | |
| Rabbit | |
| Angiogenesis, Cancer, Protein Kinase | |
| 10mM PBS (pH 7.4) and 1.0% BSA with 15mM Sodium Azide | |
| 2261 | |
| Synthetic peptide: KDLLPPAPPSSGGSRT, corresponding to amino acids 792-806 of the cytoplasmic region of Human FGFR3 with N-terminal added lysine. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| EC 2.7.10, fibroblast growth factor receptor 3, HSFGFR3EX, JTK4thanatophoric dwarfism | |
| FGFR3 | |
| IgG | |
| Affinity Purified | |
| Does not react with human FGFR1 or FGFR2. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title