missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFR3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB600-1509-0.025ml
This item is not returnable.
View return policy
Description
FGFR3 Polyclonal specifically detects FGFR3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| FGFR3 | |
| Polyclonal | |
| Unconjugated | |
| 10mM PBS (pH 7.4) and 1.0% BSA with 15mM Sodium Azide | |
| FGFR3 | |
| Synthetic peptide: KDLLPPAPPSSGGSRT, corresponding to amino acids 792-806 of the cytoplasmic region of Human FGFR3 with N-terminal added lysine. | |
| 0.025 mL | |
| Angiogenesis, Cancer, Protein Kinase | |
| 2261 | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry 1:250, Immunohistochemistry-Paraffin 1:250 | |
| EC 2.7.10, fibroblast growth factor receptor 3, HSFGFR3EX, JTK4thanatophoric dwarfism | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Does not react with human FGFR1 or FGFR2. | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction