missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFR1 Antibody (3D4F7), Novus Biologicals™
Mouse Monoclonal Antibody
£161.00
Specifications
| Antigen | FGFR1 |
|---|---|
| Clone | 3D4F7 |
| Dilution | Western Blot 1: 500-1:2000, ELISA 1:10000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
FGFR1 Monoclonal specifically detects FGFR1 in Human samples. It is validated for Western Blot, ELISA.Specifications
| FGFR1 | |
| Western Blot 1: 500-1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| 2260 | |
| Purified recombinant extracellular fragment of human FGFR1(aa33-423) fused with hIgGFc tag expressed in HEK293 cells. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 3D4F7 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Apoptosis, Core ESC Like Genes, Protein Kinase, Signal Transduction, Stem Cell Markers | |
| basic fibroblast growth factor receptor 1, BFGFR, bFGF-R-1, CD331, CD331 antigen, CEK, EC 2.7.10, EC 2.7.10.1, FGFBR, FGFR-1, fibroblast growth factor receptor 1, FLGH3, FLJ99988, FLT-2, FLT2H4, Fms-like tyrosine kinase 2, fms-related tyrosine kinase 2, H2, H5, HBGFR, heparin-binding growth factor receptor, hydroxyaryl-protein kinase, KAL 2, KAL2, N-SAM, OGD, Proto-oncogene c-Fgr, soluble FGFR1 variant 1, soluble FGFR1 variant 2 | |
| FGFR1 | |
| IgG2b | |
| Unpurified | |
| 92 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title