missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFR1 Antibody (3D4F7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-47451-0.025ml
This item is not returnable.
View return policy
Description
FGFR1 Monoclonal specifically detects FGFR1 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| FGFR1 | |
| Monoclonal | |
| Unconjugated | |
| basic fibroblast growth factor receptor 1, BFGFR, bFGF-R-1, CD331, CD331 antigen, CEK, EC 2.7.10, EC 2.7.10.1, FGFBR, FGFR-1, fibroblast growth factor receptor 1, FLGH3, FLJ99988, FLT-2, FLT2H4, Fms-like tyrosine kinase 2, fms-related tyrosine kinase 2, H2, H5, HBGFR, heparin-binding growth factor receptor, hydroxyaryl-protein kinase, KAL 2, KAL2, N-SAM, OGD, Proto-oncogene c-Fgr, soluble FGFR1 variant 1, soluble FGFR1 variant 2 | |
| Mouse | |
| 92 kDa | |
| 0.025 mL | |
| Angiogenesis, Apoptosis, Core ESC Like Genes, Protein Kinase, Signal Transduction, Stem Cell Markers | |
| 2260 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2b |
| Western Blot | |
| 3D4F7 | |
| Western Blot 1: 500-1:2000, ELISA 1:10000 | |
| FGFR1 | |
| Purified recombinant extracellular fragment of human FGFR1(aa33-423) fused with hIgGFc tag expressed in HEK293 cells. | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction