missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF basic/FGF2/bFGF Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£125.00 - £359.00
Specifications
| Antigen | FGF basic/FGF2 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1-0.2 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18208994
|
Novus Biologicals
NBP1-18579-0.025MG |
0.025 mg |
£125.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18263970
|
Novus Biologicals
NBP1-18579 |
0.2 mg |
£359.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FGF basic/FGF2/bFGF Polyclonal specifically detects FGF basic/FGF2/bFGF in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| FGF basic/FGF2 | |
| Western Blot 0.1-0.2 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Rat, Mouse | |
| P09038 | |
| 2247 | |
| Recombinant human FGF-2 (basic) | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Cancer | |
| PBS with No Preservative | |
| Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2 | |
| FGF2 | |
| IgG | |
| Protein A purified | |
| FGF-2 (basic) |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title