missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF basic/FGF2/bFGF Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-18579
This item is not returnable.
View return policy
Description
FGF basic/FGF2/bFGF Polyclonal specifically detects FGF basic/FGF2/bFGF in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| FGF basic/FGF2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with No Preservative | |
| Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| FGF-2 (basic) | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1-0.2 ug/ml | |
| P09038 | |
| FGF2 | |
| Recombinant human FGF-2 (basic) | |
| 0.2 mg | |
| Angiogenesis, Cancer | |
| 2247 | |
| Centrifuge vial prior to opening. Reconstitute in sterile water to a concentration of 0.1-1.0 mg/ml. Please note the sample size of this product will be provided in reconstituted liquid form. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction