missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse FGF-23 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
£181.00 - £403.00
Specifications
| Antigen | FGF-23 |
|---|---|
| Clone | 283511 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18731708
![]() |
R&D Systems
MAB2629-SP |
25 μg |
£181.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18111901
![]() |
R&D Systems
MAB2629 |
100 μg |
£403.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FGF-23 Monoclonal specifically detects FGF-23 in Human, Mouse samples. It is validated for Western Blot.Specifications
| FGF-23 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Human, Mouse | |
| Q9EPC2 | |
| 8074 | |
| Mouse myeloma cell line NS0-derived recombinant mouse FGF-23 Tyr25-Val251 (Arg179Gln) Accession # Q9EPC2 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human and mouse FGF-23 in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) FGF acidic, rhFGF basic, rhFGF-3, -4, -5, -6, -7, -9, -10, -11, -12, -13, -16, -17, -18, -19, -20, -21, recombinant mouse (rm) FGF-8b, rmFGF-8c, or rmFGF-15. |
| 283511 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ADHR, FGF23, FGF-23, fibroblast growth factor 23, HPDR2, HYPF, Phosphatonin, PHPTC, tumor-derived hypophosphatemia inducing factor, Tumor-derived hypophosphatemia-inducing factor | |
| FGF23 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title