missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse FGF-23 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB2629-SP
This item is not returnable.
View return policy
Description
FGF-23 Monoclonal specifically detects FGF-23 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| FGF-23 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9EPC2 | |
| FGF23 | |
| Mouse myeloma cell line NS0-derived recombinant mouse FGF-23 Tyr25-Val251 (Arg179Gln) Accession # Q9EPC2 | |
| 25 μg | |
| Primary | |
| Detects human and mouse FGF-23 in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) FGF acidic, rhFGF basic, rhFGF-3, -4, -5, -6, -7, -9, -10, -11, -12, -13, -16, -17, -18, -19, -20, -21, recombinant mouse (rm) FGF-8b, rmFGF-8c, or rmFGF-15. | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 283511 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ADHR, FGF23, FGF-23, fibroblast growth factor 23, HPDR2, HYPF, Phosphatonin, PHPTC, tumor-derived hypophosphatemia inducing factor, Tumor-derived hypophosphatemia-inducing factor | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8074 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction