Learn More
Fexofenadine hydrochloride, Thermo Scientific™
H-1 receptor antagonist
£71.65 - £109.00
Chemical Identifiers
| CAS | 153439-40-8 |
|---|---|
| Molecular Formula | C32H40ClNO4 |
| Molecular Weight (g/mol) | 538.13 |
| MDL Number | MFCD00865710 |
| InChI Key | RRJFVPUCXDGFJB-UHFFFAOYNA-N |
| Synonym | fexofenadine hydrochloride, fexofenadine hcl, allegra, telfast, 2-4-1-hydroxy-4-4-hydroxydiphenylmethyl piperidin-1-yl butyl phenyl-2-methylpropanoic acid hydrochloride, fexofenidine hydrochloride, terfenadine carboxylate hydrochloride, mdl 16455 hydrochloride, terfenidine carboxylate hydrochloride |
| PubChem CID | 63002 |
| IUPAC Name | hydrogen 2-(4-{1-hydroxy-4-[4-(hydroxydiphenylmethyl)piperidin-1-yl]butyl}phenyl)-2-methylpropanoic acid chloride |
| SMILES | [H+].[Cl-].CC(C)(C(O)=O)C1=CC=C(C=C1)C(O)CCCN1CCC(CC1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15485529
|
Thermo Scientific Alfa Aesar
J63262.MB |
25mg |
£79.80 £71.65 / 25mg Save £8.15 10% Off |
Estimated Shipment: 16-09-2026
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15495529
|
Thermo Scientific Alfa Aesar
J63262.MC |
100mg |
£109.00
100mg |
Estimated Shipment: 09-09-2026
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsFexofenadine hydrochloride is used as an antihistamine that inhibits Cox-1, Cox-2, and is a histamine H1 receptor agonist.
Solubility
Soluble in Dimethyl sulfoxide (50 mM), and methanol.
Notes
Protect from heat. Store away from oxidizing agents.
Chemical Identifiers
| 153439-40-8 | |
| 538.13 | |
| RRJFVPUCXDGFJB-UHFFFAOYNA-N | |
| 63002 | |
| [H+].[Cl-].CC(C)(C(O)=O)C1=CC=C(C=C1)C(O)CCCN1CCC(CC1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1 |
| C32H40ClNO4 | |
| MFCD00865710 | |
| fexofenadine hydrochloride, fexofenadine hcl, allegra, telfast, 2-4-1-hydroxy-4-4-hydroxydiphenylmethyl piperidin-1-yl butyl phenyl-2-methylpropanoic acid hydrochloride, fexofenidine hydrochloride, terfenadine carboxylate hydrochloride, mdl 16455 hydrochloride, terfenidine carboxylate hydrochloride | |
| hydrogen 2-(4-{1-hydroxy-4-[4-(hydroxydiphenylmethyl)piperidin-1-yl]butyl}phenyl)-2-methylpropanoic acid chloride |
Specifications
| 153439-40-8 | |
| MFCD00865710 | |
| fexofenadine hydrochloride, fexofenadine hcl, allegra, telfast, 2-4-1-hydroxy-4-4-hydroxydiphenylmethyl piperidin-1-yl butyl phenyl-2-methylpropanoic acid hydrochloride, fexofenidine hydrochloride, terfenadine carboxylate hydrochloride, mdl 16455 hydrochloride, terfenidine carboxylate hydrochloride | |
| RRJFVPUCXDGFJB-UHFFFAOYNA-N | |
| hydrogen 2-(4-{1-hydroxy-4-[4-(hydroxydiphenylmethyl)piperidin-1-yl]butyl}phenyl)-2-methylpropanoic acid chloride | |
| 63002 | |
| Fexofenadine hydrochloride |
| C32H40ClNO4 | |
| 14,4068 | |
| Soluble in Dimethyl sulfoxide (50 mM),and methanol. | |
| [H+].[Cl-].CC(C)(C(O)=O)C1=CC=C(C=C1)C(O)CCCN1CCC(CC1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 538.13 | |
| 538.12 |
Safety and Handling
missing translation for 'rtecsNumber' : CY1633377
missing translation for 'tsca' : No
Recommended Storage : Store at -20°C
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.