missing translation for 'onlineSavingsMsg'
Learn More

Fexofenadine hydrochloride, Thermo Scientific™

H-1 receptor antagonist

Brand:  Thermo Scientific Alfa Aesar J63262.MB

Quantity 25mg

 View more versions of this product

Product Code. 15485529

  • ONLINE EXCLUSIVE PRICE
    £79.80
    £71.65 / 25mg
    Save £8.15
    10% Off
 Restricted Item
Estimated Shipment: 16-09-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Fexofenadine hydrochloride is used as an antihistamine that inhibits Cox-1, Cox-2, and is a histamine H1 receptor agonist.

Solubility
Soluble in Dimethyl sulfoxide (50 mM), and methanol.

Notes
Protect from heat. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

153439-40-8
538.13
RRJFVPUCXDGFJB-UHFFFAOYNA-N
63002
[H+].[Cl-].CC(C)(C(O)=O)C1=CC=C(C=C1)C(O)CCCN1CCC(CC1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
C32H40ClNO4
MFCD00865710
fexofenadine hydrochloride, fexofenadine hcl, allegra, telfast, 2-4-1-hydroxy-4-4-hydroxydiphenylmethyl piperidin-1-yl butyl phenyl-2-methylpropanoic acid hydrochloride, fexofenidine hydrochloride, terfenadine carboxylate hydrochloride, mdl 16455 hydrochloride, terfenidine carboxylate hydrochloride
hydrogen 2-(4-{1-hydroxy-4-[4-(hydroxydiphenylmethyl)piperidin-1-yl]butyl}phenyl)-2-methylpropanoic acid chloride

Specifications

153439-40-8
MFCD00865710
14,4068
Soluble in Dimethyl sulfoxide (50 mM),and methanol.
[H+].[Cl-].CC(C)(C(O)=O)C1=CC=C(C=C1)C(O)CCCN1CCC(CC1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
538.13
538.12
C32H40ClNO4
25mg
fexofenadine hydrochloride, fexofenadine hcl, allegra, telfast, 2-4-1-hydroxy-4-4-hydroxydiphenylmethyl piperidin-1-yl butyl phenyl-2-methylpropanoic acid hydrochloride, fexofenidine hydrochloride, terfenadine carboxylate hydrochloride, mdl 16455 hydrochloride, terfenidine carboxylate hydrochloride
RRJFVPUCXDGFJB-UHFFFAOYNA-N
hydrogen 2-(4-{1-hydroxy-4-[4-(hydroxydiphenylmethyl)piperidin-1-yl]butyl}phenyl)-2-methylpropanoic acid chloride
63002
Fexofenadine hydrochloride
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.