missing translation for 'onlineSavingsMsg'
Learn More

Feruloylputrescine Trifluoroacetic Acid Salt, TRC

Product Code. 30394738
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30394738 250 mg
30407516 100 mg
30393619 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30394738 Supplier Toronto Research Chemicals Supplier No. TRCF308950250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91000-11-2
Molecular Formula C14H20N2O3•C2HF3O2
Molecular Weight (g/mol) 264.3211402
Synonym N-(4-Aminobutyl)-3-(4-hydroxy-3-methoxyphenyl)-2-propenamide Trifluoroacetic Acid Salt, N-(4-Aminobutyl)-4-hydroxy-3-methoxycinnamamide, N-Feruloylputrescine, NSC 602818 Trifluoroacetic Acid Salt, Subaphylline Trifluoroacetic Acid Salt
SMILES O=C(NCCCCN)/C=C/C1=CC=C(O)C(OC)=C1.OC(C(F)(F)F)=O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C14H20N2O3/c1-19-13-10-11(4-6-12(13)17)5-7-14(18)16-9-3-2-8-15/h4-7,10,17H,2-3,8-9,15H2,1H3,(H,16,18)/b7-5+
Chemical Name or Material Feruloylputrescine Trifluoroacetic Acid Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.