CAS RN 91000-11-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Feruloylputrescine Trifluoroacetic Acid Salt, TRC
CAS: 91000-11-2 Molecular Formula: C14H20N2O3•C2HF3O2 Molecular Weight (g/mol): 264.3211402 Synonym: N-(4-Aminobutyl)-3-(4-hydroxy-3-methoxyphenyl)-2-propenamide Trifluoroacetic Acid Salt,N-(4-Aminobutyl)-4-hydroxy-3-methoxycinnamamide,N-Feruloylputrescine,NSC 602818 Trifluoroacetic Acid Salt,Subaphylline Trifluoroacetic Acid Salt SMILES: O=C(NCCCCN)/C=C/C1=CC=C(O)C(OC)=C1.OC(C(F)(F)F)=O