missing translation for 'onlineSavingsMsg'
Learn More

Ferrocenecarboxaldehyde, 97%, Thermo Scientific Chemicals

Product Code. p-7050671
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11303658 2 g 2g
11313658 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11303658 Supplier Thermo Scientific Alfa Aesar Supplier No. 039254.04
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Ferrocenecarboxaldehyde, is used to prepare chiral ferrocene aziridinylmethanols for selective azomethine ylide cycloaddtion. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12093-10-6
Molecular Formula C11H10FeO
Molecular Weight (g/mol) 214.05
MDL Number MFCD00001429
InChI Key UQTCQJVPLIVCAX-UHFFFAOYNA-N
Synonym [Cyclopentadienyl(formylcyclopentadienyl)iron]
IUPAC Name (cyclopenta-2,4-dien-1-yl)(2-formylcyclopenta-2,4-dien-1-yl)iron
SMILES [Fe].c1cccc1.O=Cc1cccc1

Specifications

Melting Point 124°C
Quantity 2 g
Sensitivity Air sensitive
Solubility Information It is soluble in water.
Formula Weight 214.05
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material Ferrocenecarboxaldehyde

RUO – Research Use Only

Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.