CAS RN 12093-10-6
IUPAC Name:
(cyclopenta-2,4-dien-1-yl)(2-formylcyclopenta-2,4-dien-1-yl)iron
Synonyms:
cyclopenta-2,4-dien-1-yl(2-formylcyclopenta-2,4-dien-1-yl)iron
Molecular Weight (g/mol):
214.05
Molecular Formula:
C11H10FeO
InChi Key:
UQTCQJVPLIVCAX-UHFFFAOYNA-N
SMILES:
O=CC1=CC=CC1[Fe]C1C=CC=C1
1
–
1
of
1
results
1
Ferrocenecarboxaldehyde, 97%, Thermo Scientific Chemicals
CAS: 12093-10-6 Molecular Formula: C11H10FeO Molecular Weight (g/mol): 214.05 MDL Number: MFCD00001429 InChI Key: UQTCQJVPLIVCAX-UHFFFAOYNA-N Synonym: [Cyclopentadienyl(formylcyclopentadienyl)iron] IUPAC Name: (cyclopenta-2,4-dien-1-yl)(2-formylcyclopenta-2,4-dien-1-yl)iron SMILES: [Fe].c1cccc1.O=Cc1cccc1