missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ferritin Light Chain Monoclonal Antibody (H19), Invitrogen™

Mouse Monoclonal Antibody
£178.00 - £386.00
Specifications
| Antigen | Ferritin Light Chain |
|---|---|
| Clone | H19 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17624286
|
Invitrogen™
MA543828 |
20 μL |
£178.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17614286
![]() |
Invitrogen™
MA543827 |
100 μL |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes the light subunit of the ferritin protein. Ferritin is the major intracellular iron storage protein in prokaryotes and eukaryotes. It is composed of 24 subunits of the heavy and light ferritin chains. Variation in ferritin subunit composition may affect the rates of iron uptake and release in different tissues. A major function of ferritin is the storage of iron in a soluble and nontoxic state. Defects in this light chain ferritin gene are associated with several neurodegenerative diseases and hyperferritinemia-cataract syndrome. This gene has multiple pseudogenes.Specifications
| Ferritin Light Chain | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| ferritin L subunit; Ferritin L subunit 1; ferritin L-chain; ferritin light chain; Ferritin light chain 1; ferritin light polypeptide-like 3; ferritin, light polypeptide; fril; Ftl; Ftl1; Ftl-1; L-ferritin; LFTD; NBIA3 | |
| FTL, Ftl1 | |
| IgG2a κ | |
| Protein A/G | |
| Antibody |
| H19 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| P02792, P02793 | |
| 2512, 29292 | |
| Ser2-Asp175 (Accession P02792), with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| FTL |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title